Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromoisophthalic acid, 95%

Manufacturer:  Alfa Aesar™ H2687406

Quantity 5g

 View more versions of this product

Catalog No. AAH2687406


  Check Availability
Add to cart

Specifications

Description

Applications
5-Bromoisophthalic acid, is used as a pharmaceutical intermediate. It is also used in the polymer industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Description & Specifications

Chemical Identifiers

23351-91-9
245.03
5-Bromoisophthalic acid
C8H5BrO4
MFCD00152019

Specifications

23351-91-9
5g
C8H5BrO4
MFCD00152019
95%
280° to 286°C
Odorless
245.03
5-Bromoisophthalic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit