Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoisoquinoline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
34784-04-8
C9H6BrN
208.058
MFCD01646405
CYJZJGYYTFQQBY-UHFFFAOYSA-N
isoquinoline, 5-bromo,5-bromo-isoquinoline,5-bromo isoquinoline,5-bromoisoquinoline?,pubchem6214,acmc-1adiy,5-bromoisoquinoline,ksc226k2l,buttpark 44\07-76,5-bromoisoquinoline 250mg
736487
5-bromoisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)Br
34784-04-8
C9H6BrN
208.058
MFCD01646405
CYJZJGYYTFQQBY-UHFFFAOYSA-N
isoquinoline, 5-bromo,5-bromo-isoquinoline,5-bromo isoquinoline,5-bromoisoquinoline?,pubchem6214,acmc-1adiy,5-bromoisoquinoline,ksc226k2l,buttpark 44\07-76,5-bromoisoquinoline 250mg
736487
5-bromoisoquinoline
C1=CC2=C(C=CN=C2)C(=C1)Br
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 208.06
Chemical Name or Material 5-Bromoisoquinoline

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning