Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(bromomethyl)-3-methylisoxazole, 95%, Thermo Scientific™

Catalog Number cc27008cb
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
36958-61-9
C5H6BrNO
176.013
MFCD09879901
DMUJLHLWCUMHRT-UHFFFAOYSA-N
5-bromomethyl-3-methylisoxazole, 5-bromomethyl-3-methyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-methyl, 3-methyl-5-bromomethylisoxazole
10607354
5-(bromomethyl)-3-methyl-1,2-oxazole
CC1=NOC(=C1)CBr
36958-61-9
C5H6BrNO
176.013
MFCD09879901
DMUJLHLWCUMHRT-UHFFFAOYSA-N
5-bromomethyl-3-methylisoxazole, 5-bromomethyl-3-methyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-methyl, 3-methyl-5-bromomethylisoxazole
10607354
5-(bromomethyl)-3-methyl-1,2-oxazole
CC1=NOC(=C1)CBr
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.01
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-3-methylisoxazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit