Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromonicotinic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
20826-04-4
C6H4BrNO2
202.01
MFCD00009783
FQIUCPGDKPXSLL-UHFFFAOYSA-N
5-bromonicotinic acid,5-bromo nicotinic acid,5-bromo-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 5-bromo,nicotinic acid, 5-bromo,5-bromo-nicotinic acid,3-bromo-5-pyridine carboxylic acid,3-bromo pyridine-5-carboxylic acid,pubchem1211,acmc-209fev
88707
5-bromopyridine-3-carboxylic acid
C1=C(C=NC=C1Br)C(=O)O
20826-04-4
C6H4BrNO2
202.01
MFCD00009783
FQIUCPGDKPXSLL-UHFFFAOYSA-N
5-bromonicotinic acid,5-bromo nicotinic acid,5-bromo-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 5-bromo,nicotinic acid, 5-bromo,5-bromo-nicotinic acid,3-bromo-5-pyridine carboxylic acid,3-bromo pyridine-5-carboxylic acid,pubchem1211,acmc-209fev
88707
5-bromopyridine-3-carboxylic acid
C1=C(C=NC=C1Br)C(=O)O
Melting Point 178°C to 180°C
Infrared Spectrum Authentic
Color Brown or Gray-Yellow to White
Quantity 50g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula Br(C5H3N)COOH
Beilstein 22, 44
Formula Weight 202.01
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 5-Bromonicotinic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-065-5