Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromopyrazine-2-carboxylic acid, 95%

Click to view available options
Quantity:
250mg
1g
876161-05-6
C5H3BrN2O2
202.995
MFCD09909672
NPJHCKULJAIWGV-UHFFFAOYSA-N
5-bromo-2-pyrazinecarboxylic acid,2-pyrazinecarboxylicacid, 5-bromo,2-pyrazinecarboxylic acid, 5-bromo,5-bromo-2-pyrazinecarboxylicacid,ksc446e0j,2-pyrazinecarboxylicacid,5-bromo,2-bromopyrazine-5-carboxylic acid
18779790
5-bromopyrazine-2-carboxylic acid
C1=C(N=CC(=N1)Br)C(=O)O
876161-05-6
C5H3BrN2O2
202.995
MFCD09909672
NPJHCKULJAIWGV-UHFFFAOYSA-N
5-bromo-2-pyrazinecarboxylic acid,2-pyrazinecarboxylicacid, 5-bromo,2-pyrazinecarboxylic acid, 5-bromo,5-bromo-2-pyrazinecarboxylicacid,ksc446e0j,2-pyrazinecarboxylicacid,5-bromo,2-bromopyrazine-5-carboxylic acid
18779790
5-bromopyrazine-2-carboxylic acid
C1=C(N=CC(=N1)Br)C(=O)O
Quantity 1g
Formula Weight 203
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Bromopyrazine-2-carboxylic acid

storageNote1 : -30° to -10°C