Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4595-59-9
C4H3BrN2
158.99
MFCD00006117
GYCPLYCTMDTEPU-UHFFFAOYSA-N
pyrimidine, 5-bromo,5-bromo pyrimidine,5-bromo-1,3-diazine,5-bromo-pyrimidine,5-brpy compound,5-brompyrimidin,5-brornopyrimidine,zlchem 349,pubchem6902,11-cis-retinyl-palmitate
78344
5-bromopyrimidine
C1=C(C=NC=N1)Br
4595-59-9
C4H3BrN2
158.99
MFCD00006117
GYCPLYCTMDTEPU-UHFFFAOYSA-N
pyrimidine, 5-bromo,5-bromo pyrimidine,5-bromo-1,3-diazine,5-bromo-pyrimidine,5-brpy compound,5-brompyrimidin,5-brornopyrimidine,zlchem 349,pubchem6902,11-cis-retinyl-palmitate
78344
5-bromopyrimidine
C1=C(C=NC=N1)Br
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Color Undesignated
Melting Point 69.0°C to 73.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H3N2)Br
Formula Weight 158.99
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material 5-Bromopyrimidine, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 224-992-1

TSCA : TSCA