Alfa Aesar™ 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one, 99%

Manufacturer: Alfa Aesar™ H50582MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50582MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75822-54-7
    Molecular Formula C10H8BrNO3
    Formula Weight 270.08
    MDL Number MFCD00097597
    Synonym 5-Bromoisatin 3-(ethylene acetal)
    Chemical Name or Material 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one

    Specifications

    Formula Weight 270.08
    Molecular Formula C10H8BrNO3
    MDL Number MFCD00097597
    CAS 75822-54-7
    Synonym 5-Bromoisatin 3-(ethylene acetal)
    Quantity 250mg
    RTECS Number WH1306502
    Chemical Name or Material 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit