Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromothiazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3034-55-7
C3H2BrNS
164.02
MFCD07787394
DWUPYMSVAPQXMS-UHFFFAOYSA-N
5-bromothiazole,thiazole, 5-bromo,5-bromo-thiazole,5-bromthiazole,thiazole,5-bromo,5-bromothiazol,5-bromo thiazole,zlchem 497,pubchem2077,2-bromo-1,4-thiazole
546059
5-bromo-1,3-thiazole
C1=C(SC=N1)Br
3034-55-7
C3H2BrNS
164.02
MFCD07787394
DWUPYMSVAPQXMS-UHFFFAOYSA-N
5-bromothiazole,thiazole, 5-bromo,5-bromo-thiazole,5-bromthiazole,thiazole,5-bromo,5-bromothiazol,5-bromo thiazole,zlchem 497,pubchem2077,2-bromo-1,4-thiazole
546059
5-bromo-1,3-thiazole
C1=C(SC=N1)Br
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Flash Point 76.7°C
Color Yellow to Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 164.02
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 5-Bromothiazole
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning