Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromothiophene-2-carboxaldoxime, 97%

Click to view available options
Quantity:
1g
5g
2160-63-6
C5H4BrNOS
206.057
MFCD00126375
URLSTLFAZRSULI-XVNBXDOJSA-N
5-bromothiophene-2-carboxaldehyde oxime,5-bromothiophene-2-carbaldehyde oxime,5-bromo-2-thiophenaldoxime,5-bromothiophene-2-carboxaldoxime,5-bromo-2-thiophenecarboxaldehyde oxime,2-thiophenecarboxaldehyde, 5-bromo-, oxime,5-bromo-thiophene-2-carbaldehyde oxime,5-bromo 2-thienyl hydroxyimino methane,2e-5-bromo-2-thiophenecarboxaldehyde oxime,e-1-5-bromothiophen-2-yl-n-hydroxymethanimine
9578377
(NE)-N-[(5-bromothiophen-2-yl)methylidene]hydroxylamine
C1=C(SC(=C1)Br)C=NO
2160-63-6
C5H4BrNOS
206.057
MFCD00126375
URLSTLFAZRSULI-XVNBXDOJSA-N
5-bromothiophene-2-carboxaldehyde oxime,5-bromothiophene-2-carbaldehyde oxime,5-bromo-2-thiophenaldoxime,5-bromothiophene-2-carboxaldoxime,5-bromo-2-thiophenecarboxaldehyde oxime,2-thiophenecarboxaldehyde, 5-bromo-, oxime,5-bromo-thiophene-2-carbaldehyde oxime,5-bromo 2-thienyl hydroxyimino methane,2e-5-bromo-2-thiophenecarboxaldehyde oxime,e-1-5-bromothiophen-2-yl-n-hydroxymethanimine
9578377
(NE)-N-[(5-bromothiophen-2-yl)methylidene]hydroxylamine
C1=C(SC(=C1)Br)C=NO
Quantity 1g
Melting Point 132°C to 134°C
Formula Weight 206.06
Percent Purity 97%
Chemical Name or Material 5-Bromothiophene-2-carboxaldoxime

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning