Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromothiophenesulfonyl chloride, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
55854-46-1
C4H2BrClO2S2
261.532
MFCD00084909
WGYBIEOLAFYDEC-UHFFFAOYSA-N
5-bromothiophenesulfonyl chloride,5-bromo-2-thiophenesulfonyl chloride,5-bromo-thiophene-2-sulfonyl chloride,5-bromothiophene-2-sulphonyl chloride,5-bromo-2-chlorosulphonyl thiophene,5-bromo-2-chlorosulfonyl thiophene,5-bromothiophenesulphonyl chloride,buttpark 21\07-09,5-bromo 2-thienyl chlorosulfone,5-bromothiophen-2-sulfonyl chloride
2733924
5-bromothiophene-2-sulfonyl chloride
C1=C(SC(=C1)Br)S(=O)(=O)Cl
55854-46-1
C4H2BrClO2S2
261.532
MFCD00084909
WGYBIEOLAFYDEC-UHFFFAOYSA-N
5-bromothiophenesulfonyl chloride,5-bromo-2-thiophenesulfonyl chloride,5-bromo-thiophene-2-sulfonyl chloride,5-bromothiophene-2-sulphonyl chloride,5-bromo-2-chlorosulphonyl thiophene,5-bromo-2-chlorosulfonyl thiophene,5-bromothiophenesulphonyl chloride,buttpark 21\07-09,5-bromo 2-thienyl chlorosulfone,5-bromothiophen-2-sulfonyl chloride
2733924
5-bromothiophene-2-sulfonyl chloride
C1=C(SC(=C1)Br)S(=O)(=O)Cl
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 261.55
Physical Form Crystalline Low Melting Solid
Percent Purity ≥95% (ex Cl)
Chemical Name or Material 5-Bromothiophenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage