Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Carbethoxyuracil, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
28485-17-8
C7H8N2O4
184.15
MFCD00057337
MKNYHTGOVKPZMU-UHFFFAOYSA-N
5-carbethoxyuracil,ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate,ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate,isoorotic acid ethyl ester,ethyl 2,4-dihydroxypyrimidine-5-carboxylate,ethyl uracil-5-carboxylate,ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate,5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester,ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate,pubchem13254
101411
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CNC(=O)NC1=O
28485-17-8
C7H8N2O4
184.15
MFCD00057337
MKNYHTGOVKPZMU-UHFFFAOYSA-N
5-carbethoxyuracil,ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate,ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate,isoorotic acid ethyl ester,ethyl 2,4-dihydroxypyrimidine-5-carboxylate,ethyl uracil-5-carboxylate,ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate,5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester,ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate,pubchem13254
101411
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate
CCOC(=O)C1=CNC(=O)NC1=O
Packaging Glass bottle
Melting Point 232.0°C to 235.0°C
Color White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 184.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Carbethoxyuracil
Show More Show Less

EINECSNumber : 249-053-3