Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Carbethoxyuracil, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Specifications
Color | White |
Melting Point | 232.0°C to 235.0°C |
Quantity | 25g |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (HPLC) |
Formula Weight | 184.15 |
Physical Form | Crystalline Powder |
Show More |
Safety and Handling
EINECSNumber : 249-053-3