Learn More
Aat BioquestSupplier Diversity Partner 5-Carboxy-4'-aminomethylfluorescein DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 207
5-Carboxy-4'-aminomethylfluorescein is a key building block for developing Big Dye DNA sequencing terminators. It can also be incorporated into a peptide sequence to give fluoresceinated peptides.
Catalog No. 50-195-2811