Alfa Aesar™ 5-Chloro-1,3-benzodioxole, 98%

Manufacturer: Alfa Aesar™ B2253430

Quantity 250g

 View more versions of this product

Catalog No. AAB2253430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7228-38-8
    Molecular Formula C7H5ClO2
    Formula Weight 156.57
    MDL Number MFCD00010842
    Synonym 3,4-Methylenedioxychlorobenzene
    Chemical Name or Material 5-Chloro-1,3-benzodioxole

    Specifications

    Formula Weight 156.57
    Molecular Formula C7H5ClO2
    MDL Number MFCD00010842
    CAS 7228-38-8
    Synonym 3,4-Methylenedioxychlorobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 93°C (201°F)
    Boiling Point 185° to 187°C
    Quantity 250g
    Chemical Name or Material 5-Chloro-1,3-benzodioxole
    Assay 98%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit