Alfa Aesar™ 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

Manufacturer: Alfa Aesar™ H50546MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50546MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88398-46-3
    Molecular Formula C5H8ClN3O2S
    Formula Weight 209.66
    MDL Number MFCD00105874
    Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

    Specifications

    Formula Weight 209.66
    Molecular Formula C5H8ClN3O2S
    MDL Number MFCD00105874
    CAS 88398-46-3
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 250mg
    Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

    Description

    Applications
    5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit