Alfa Aesar™ 5-Chloro-1-phenyl-1H-tetrazole, 98+%

Manufacturer: Alfa Aesar™ B2227218

Quantity 50g

 View more versions of this product

Catalog No. AAB2227218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14210-25-4
    Molecular Formula C7H5ClN4
    Formula Weight 180.6
    MDL Number MFCD00003128
    UN Number UN1325
    Beilstein 139070
    Chemical Name or Material 5-Chloro-1-phenyl-1H-tetrazole

    Specifications

    Formula Weight 180.6
    Melting Point 122° to 124°C
    Quantity 50g
    TSCA TSCA
    Molecular Formula C7H5ClN4
    MDL Number MFCD00003128
    CAS 14210-25-4
    UN Number UN1325
    Beilstein 139070
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 5-Chloro-1-phenyl-1H-tetrazole
    Assay ≥98%

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit