Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole, 95%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
1030574-44-7
MFCD15173053
4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline
1030574-44-7
MFCD15173053
4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline
Quantity 5g
Beilstein 22393779
Formula Weight 271.75
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole