Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-cyanobenzeneboronic acid, 98%

Click to view available options
Quantity:
250mg
1g
1072946-52-1
C7H5BClNO2
181.382
JXIINQQPEHZQAY-UHFFFAOYSA-N
5-chloro-2-cyanophenyl boronic acid,5-chloro-2-cyanophenyl boronicacid,boronicacid,b-5-chloro-2-cyanophenyl
46738934
(5-chloro-2-cyanophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)C#N)(O)O
1072946-52-1
C7H5BClNO2
181.382
JXIINQQPEHZQAY-UHFFFAOYSA-N
5-chloro-2-cyanophenyl boronic acid,5-chloro-2-cyanophenyl boronicacid,boronicacid,b-5-chloro-2-cyanophenyl
46738934
(5-chloro-2-cyanophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)C#N)(O)O
Quantity 1g
Formula Weight 225.84
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-cyanobenzeneboronic acid

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures