Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-fluorobenzamide, 97+%

Click to view available options
Quantity:
2g
10g
261762-57-6
C7H5ClFNO
173.571
MFCD01631435
VTSILFLCXIRODM-UHFFFAOYSA-N
2-fluoro-5-chlorobenzamide,pubchem9136,5-chloranyl-2-fluoranyl-benzamide,5-chloro-2-fluorobenzamide,97+%
2773592
5-chloro-2-fluorobenzamide
C1=CC(=C(C=C1Cl)C(=O)N)F
261762-57-6
C7H5ClFNO
173.571
MFCD01631435
VTSILFLCXIRODM-UHFFFAOYSA-N
2-fluoro-5-chlorobenzamide,pubchem9136,5-chloranyl-2-fluoranyl-benzamide,5-chloro-2-fluorobenzamide,97+%
2773592
5-chloro-2-fluorobenzamide
C1=CC(=C(C=C1Cl)C(=O)N)F
Quantity 2g
Melting Point 134°C to 138°C
Formula Weight 173.58
Percent Purity ≥97%
Chemical Name or Material 5-Chloro-2-fluorobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning