Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-fluorobenzoic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
394-30-9
C7H4ClFO2
174.555
MFCD00665762
WGAVMKXCDMQVNF-UHFFFAOYSA-N
2-fluoro-5-chlorobenzoic acid,5-chloro-2-fluoro-benzoic acid,5-chloro-2-fluorobenzoicacid,benzoic acid, 5-chloro-2-fluoro,pubchem1375,acmc-1aigz,ksc223m7h,rarechem al bo 0944,5-chloro-2-fluorobenzoic acid,benzoicacid, 5-chloro-2-fluoro
2736537
5-chloro-2-fluorobenzoic acid
C1=CC(=C(C=C1Cl)C(=O)O)F
394-30-9
C7H4ClFO2
174.555
MFCD00665762
WGAVMKXCDMQVNF-UHFFFAOYSA-N
2-fluoro-5-chlorobenzoic acid,5-chloro-2-fluoro-benzoic acid,5-chloro-2-fluorobenzoicacid,benzoic acid, 5-chloro-2-fluoro,pubchem1375,acmc-1aigz,ksc223m7h,rarechem al bo 0944,5-chloro-2-fluorobenzoic acid,benzoicacid, 5-chloro-2-fluoro
2736537
5-chloro-2-fluorobenzoic acid
C1=CC(=C(C=C1Cl)C(=O)O)F
Quantity 1g
Melting Point 148°C to 150°C
Beilstein 2614286
Formula Weight 174.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning