Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-fluorobenzonitrile, 97+%

Click to view available options
Quantity:
1g
5g
57381-34-7
C7H3ClFN
155.556
MFCD01631434
GJNJDELEHIGPKJ-UHFFFAOYSA-N
2-fluoro-5-chlorobenzonitrile,5-chloro-2-fluorobenzonitirle,5-chloro-2-fluorocyanobenzene,benzonitrile, 5-chloro-2-fluoro,3-chloro-6-fluorobenzonitrile,5-chloro-2-fluoro-benzonitrile,5-chloro-2-fluorobenzenecarbonitrile,ncr cg ff,pubchem1581,acmc-209lxs
93653
5-chloro-2-fluorobenzonitrile
C1=CC(=C(C=C1Cl)C#N)F
57381-34-7
C7H3ClFN
155.556
MFCD01631434
GJNJDELEHIGPKJ-UHFFFAOYSA-N
2-fluoro-5-chlorobenzonitrile,5-chloro-2-fluorobenzonitirle,5-chloro-2-fluorocyanobenzene,benzonitrile, 5-chloro-2-fluoro,3-chloro-6-fluorobenzonitrile,5-chloro-2-fluoro-benzonitrile,5-chloro-2-fluorobenzenecarbonitrile,ncr cg ff,pubchem1581,acmc-209lxs
93653
5-chloro-2-fluorobenzonitrile
C1=CC(=C(C=C1Cl)C#N)F
Melting Point 66°C to 69°C
Quantity 1g
UN Number UN3439
Formula Weight 155.56
Percent Purity ≥97%
Chemical Name or Material 5-Chloro-2-fluorobenzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.