Alfa Aesar™ 5-Chloro-2-fluorobenzotrifluoride, 98+%

Manufacturer: Alfa Aesar™ H2620803

Quantity 1g

 View more versions of this product

Catalog No. AAH2620803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89634-74-2
    Molecular Formula C7H3ClF4
    Formula Weight 198.55
    MDL Number MFCD00060655
    UN Number UN1993
    Chemical Name or Material 5-Chloro-2-fluorobenzotrifluoride

    Specifications

    Formula Weight 198.55
    Molecular Formula C7H3ClF4
    MDL Number MFCD00060655
    UN Number UN1993
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 47°C (117°F)
    Quantity 1g
    CAS 89634-74-2
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Boiling Point 145°C
    Chemical Name or Material 5-Chloro-2-fluorobenzotrifluoride
    Assay ≥98%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit