Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-fluorobenzoyl chloride, 97%
Click to view available options
Quantity:
1g
5g
Description
Applications
2-Bromo-5-fluorobenzoic acid is used as pharmaceutical intermediates.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
2-Bromo-5-fluorobenzoic acid is used as pharmaceutical intermediates.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
Specifications
Sensitivity | Moisture sensitive |
Refractive Index | 1.553 |
Density | 1.459 |
Boiling Point | 103°C to 104°C (15mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 1g |
UN Number | UN3265 |
Beilstein | 3247304 |
Formula Weight | 193.01 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures