Alfa Aesar™ 5-Chloro-2-fluoropyridine, 97%

Manufacturer: Alfa Aesar™ H2694514

Quantity 25g

 View more versions of this product

Catalog No. AAH2694514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1480-65-5
    Molecular Formula C5H3ClFN
    Formula Weight 131.54
    MDL Number MFCD04112513
    UN Number UN2924
    Chemical Name or Material 5-Chloro-2-fluoropyridine

    Specifications

    Formula Weight 131.54
    MDL Number MFCD04112513
    UN Number UN2924
    Odor Unpleasant
    Molecular Formula C5H3ClFN
    CAS 1480-65-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H226-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Flash Point 52°C (126°F)
    Boiling Point 149°C
    Quantity 25g
    Chemical Name or Material 5-Chloro-2-fluoropyridine
    Assay 97%

    GHS H Statement
    H314-H318-H226-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit