Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-fluoropyridine, 97%

Manufacturer:  Alfa Aesar™ H2694506

Quantity 5g

 View more versions of this product

Catalog No. AAH2694506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1480-65-5
131.54
UN2924
C5H3ClFN
MFCD04112513
5-Chloro-2-fluoropyridine

Specifications

52°C (126°F)
149°C
1480-65-5
131.54
UN2924
97%
Unpleasant
5g
C5H3ClFN
MFCD04112513
5-Chloro-2-fluoropyridine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit