Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5'-Chloro-2'-hydroxyacetophenone, 99%

Click to view available options
Quantity:
1g
5g
1450-74-4
C8H7ClO2
170.592
MFCD00067788
XTGCUDZCCIRWHL-UHFFFAOYSA-N
5'-chloro-2'-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethanone,5-chloro-2-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethan-1-one,2-acetyl-4-chlorophenol,ethanone, 1-5-chloro-2-hydroxyphenyl,2'-hydroxy-5'-chloroacetophenone,acetophenone, 5'-chloro-2'-hydroxy,2-hydroxy-5-chloro acetophenone,1-5-chloro-2-hydroxyphenyl-1-ethanone
74061
1-(5-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)O
1450-74-4
C8H7ClO2
170.592
MFCD00067788
XTGCUDZCCIRWHL-UHFFFAOYSA-N
5'-chloro-2'-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethanone,5-chloro-2-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethan-1-one,2-acetyl-4-chlorophenol,ethanone, 1-5-chloro-2-hydroxyphenyl,2'-hydroxy-5'-chloroacetophenone,acetophenone, 5'-chloro-2'-hydroxy,2-hydroxy-5-chloro acetophenone,1-5-chloro-2-hydroxyphenyl-1-ethanone
74061
1-(5-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)O
Boiling Point 107°C to 109°C (12mmHg)
Melting Point 52°C to 53°C
Quantity 1g
Beilstein 1365589
Formula Weight 170.6
Percent Purity 99%
Chemical Name or Material 5'-Chloro-2'-hydroxyacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-916-8

TSCA : No

Recommended Storage : Ambient temperatures