Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-iodopyridine, 98%

Click to view available options
Quantity:
1g
5g
244221-57-6
C5H3ClIN
239.44
MFCD07368864
CXWLXKZIXLOBCC-UHFFFAOYSA-N
2-iodo-5-chloropyridine,5-chloro-2-iodo-pyridine,pubchem6574,acmc-209gc5,ksc493k7f,pyridine, 5-chloro-2-iodo
11118148
5-chloro-2-iodopyridine
C1=CC(=NC=C1Cl)I
244221-57-6
C5H3ClIN
239.44
MFCD07368864
CXWLXKZIXLOBCC-UHFFFAOYSA-N
2-iodo-5-chloropyridine,5-chloro-2-iodo-pyridine,pubchem6574,acmc-209gc5,ksc493k7f,pyridine, 5-chloro-2-iodo
11118148
5-chloro-2-iodopyridine
C1=CC(=NC=C1Cl)I
Applications
It is applied in an efficient preparation of indolizines through a tandem palladium-catalyzed cross-coupling reaction and cycloisomerization and also in the synthesis, and structure-activity relationships of 2-aminoquinazoline derivatives as a novel class of metabotropic glutamate receptor 5 negative allosteric modulators.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Keep it refrigerated.
Sensitivity Light Sensitive
Quantity 1g
Beilstein 9321405
Formula Weight 239.44
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-iodopyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C