Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-iodotoluene, 97%

Click to view available options
Quantity:
1g
5g
23399-70-4
C7H6ClI
252.479
MFCD00060665
ZZNUQVQAJVZRKW-UHFFFAOYSA-N
5-chloro-2-iodotoluene,benzene, 4-chloro-1-iodo-2-methyl,4-chloro-1-iodo-2-methyl-benzene,4-chloro-2-methyl-iodobenzene,acmc-20aich,2-iodo-5-chlorotoluene,4-chloro-2-methyliodobenzene,ksc493k6n,1-iodo-2-methyl-4-chlorobenzene,benzene, 4-chloro-2-methyl-1-iodo
2735744
4-chloro-1-iodo-2-methylbenzene
CC1=C(C=CC(=C1)Cl)I
23399-70-4
C7H6ClI
252.479
MFCD00060665
ZZNUQVQAJVZRKW-UHFFFAOYSA-N
5-chloro-2-iodotoluene,benzene, 4-chloro-1-iodo-2-methyl,4-chloro-1-iodo-2-methyl-benzene,4-chloro-2-methyl-iodobenzene,acmc-20aich,2-iodo-5-chlorotoluene,4-chloro-2-methyliodobenzene,ksc493k6n,1-iodo-2-methyl-4-chlorobenzene,benzene, 4-chloro-2-methyl-1-iodo
2735744
4-chloro-1-iodo-2-methylbenzene
CC1=C(C=CC(=C1)Cl)I
Boiling Point 120°C to 122°C (15mmHg)
Quantity 1g
Formula Weight 252.48
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-iodotoluene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning