Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Chloro-2-methoxyphenyl isocyanate, 97%
Click to view available options
Quantity:
10g
Specifications
Sensitivity | Moisture sensitive |
Quantity | 10g |
Melting Point | 58°C to 62°C |
UN Number | UN2811 |
Beilstein | 2719682 |
Formula Weight | 183.6 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-methoxyphenyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
H302+H332-H315-H319-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures