Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxypyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
13473-01-3
C6H6ClNO
143.57
NPYYXUYLIHZYOU-UHFFFAOYSA-N
5-chloro-2-methoxy-pyridine,pyridine, 5-chloro-2-methoxy,2-methoxy-5-chloro pyridine,pubchem6602,2-methoxy-5-chloropyridine,acmc-1c02r,ksc494g7j,5-chloranyl-2-methoxy-pyridine,5-chloro-2-methoxypyridine
4738276
5-chloro-2-methoxypyridine
COC1=NC=C(C=C1)Cl
13473-01-3
C6H6ClNO
143.57
NPYYXUYLIHZYOU-UHFFFAOYSA-N
5-chloro-2-methoxy-pyridine,pyridine, 5-chloro-2-methoxy,2-methoxy-5-chloro pyridine,pubchem6602,2-methoxy-5-chloropyridine,acmc-1c02r,ksc494g7j,5-chloranyl-2-methoxy-pyridine,5-chloro-2-methoxypyridine
4738276
5-chloro-2-methoxypyridine
COC1=NC=C(C=C1)Cl
Flash Point 53°C
Packaging Glass Bottle
Boiling Point 180°C to 184°C
Density 1.1930g/mL
Specific Gravity 1.193
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 143.57
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxypyridine

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning