Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Chloro-2-methoxypyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Density | 1.1930g/mL |
Boiling Point | 180°C to 184°C |
Flash Point | 53°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Gravity | 1.193 |
Formula Weight | 143.57 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-methoxypyridine |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Flammable liquid and vapour. Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rin |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction