Alfa Aesar™ 5-Chloro-2-methylbenzothiazole, 98%

Manufacturer: Alfa Aesar™ H5574303

Quantity 1g

 View more versions of this product

Catalog No. AAH5574303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1006-99-1
    Formula Weight 183.66
    MDL Number MFCD00022881
    Chemical Name or Material 5-Chloro-2-methylbenzothiazole

    Specifications

    Formula Weight 183.66
    Melting Point 65° to 70°C
    Physical Form Solid
    MDL Number MFCD00022881
    CAS 1006-99-1
    Color White to pale brown
    Quantity 1g
    TSCA TSCA
    RTECS Number DL3325000
    Chemical Name or Material 5-Chloro-2-methylbenzothiazole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit