Alfa Aesar™ 5-Chloro-2-methylphenol, 98%

Manufacturer: Alfa Aesar™ B2552714

Quantity 25g

 View more versions of this product

Catalog No. AAB2552714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5306-98-9
    Molecular Formula C7H7ClO
    Formula Weight 142.59
    MDL Number MFCD00060672
    UN Number UN3437
    Beilstein 1906683
    Synonym 5-Chloro-o-cresol
    Chemical Name or Material 5-Chloro-2-methylphenol

    Specifications

    Formula Weight 142.59
    Molecular Formula C7H7ClO
    Synonym 5-Chloro-o-cresol
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Boiling Point 109° to 112°C (15mmHg)
    Melting Point 70° to 75°C
    Quantity 25g
    MDL Number MFCD00060672
    UN Number UN3437
    Beilstein 1906683
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 5306-98-9
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
    Chemical Name or Material 5-Chloro-2-methylphenol
    Assay 98%

    GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit