Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-methylphenyl isothiocyanate, 97%

Click to view available options
Quantity:
5g
25g
100g
19241-36-2
C8H6ClNS
183.653
MFCD00046801
VDBDGAPNWNWUSA-UHFFFAOYSA-N
5-chloro-2-methylphenyl isothiocyanate,isothiocyanic acid, 5-chloro-o-tolyl ester,5-chloro-o-tolyl isothiocyanate,5-chloro-2-methylphenylisothiocyanate,benzene,4-chloro-2-isothiocyanato-1-methyl,acmc-20am4h,3-12-00-01913 beilstein handbook reference,4-chloro-2-isothiocyanatotoluene,5-chloro-2-methylbenzenisothiocyanate,5-chloro-2-methylphenyl-isothiocyanate
29519
4-chloro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)Cl)N=C=S
19241-36-2
C8H6ClNS
183.653
MFCD00046801
VDBDGAPNWNWUSA-UHFFFAOYSA-N
5-chloro-2-methylphenyl isothiocyanate,isothiocyanic acid, 5-chloro-o-tolyl ester,5-chloro-o-tolyl isothiocyanate,5-chloro-2-methylphenylisothiocyanate,benzene,4-chloro-2-isothiocyanato-1-methyl,acmc-20am4h,3-12-00-01913 beilstein handbook reference,4-chloro-2-isothiocyanatotoluene,5-chloro-2-methylbenzenisothiocyanate,5-chloro-2-methylphenyl-isothiocyanate
29519
4-chloro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)Cl)N=C=S
Boiling Point 268°C to 272°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5g
UN Number UN2922
Beilstein 2717659
Formula Weight 183.66
Percent Purity 97%
Density 1.248
Refractive Index 1.646
Sensitivity Moisture sensitive
Chemical Name or Material 5-Chloro-2-methylphenyl isothiocyanate
Show More Show Less

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

RTECSNumber : NX8476000

TSCA : No

Recommended Storage : Ambient temperatures