Alfa Aesar™ 5-Chloro-2-nitroaniline, 97%

Manufacturer: Alfa Aesar™ A1124106

Quantity 5g

 View more versions of this product

Catalog No. AAA1124106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1635-61-6
    Molecular Formula C6H5ClN2O2
    Formula Weight 172.57
    MDL Number MFCD00007776
    UN Number UN2237
    Beilstein 2210201
    Chemical Name or Material 5-Chloro-2-nitroaniline

    Specifications

    Formula Weight 172.57
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Weak, characteristic
    Molecular Formula C6H5ClN2O2
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Melting Point 124° to 128°C
    Quantity 5g
    MDL Number MFCD00007776
    CAS 1635-61-6
    Beilstein 2210201
    Health Hazard 1 Danger
    UN Number UN2237
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES
    Chemical Name or Material 5-Chloro-2-nitroaniline
    Assay 97%

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit