Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-nitrodiphenylamine, 98%

Click to view available options
Quantity:
5g
25781-92-4
C12H9ClN2O2
248.666
MFCD00007287
FPKHZBVGKMTUHB-UHFFFAOYSA-N
5-chloro-2-nitrodiphenylamine,benzenamine, 5-chloro-2-nitro-n-phenyl,5-chloro-2-nitrophenyl phenylamine,ghl.pd_mitscher_leg0.920,ksc203m3b,2-anilino-4-chloronitrobenzene,fpkhzbvgkmtuhb-uhfffaoysa,5-chloro-2-nitro-n-phenyl-benzenamine,benzenamine,5-chloro-2-nitro-n-phenyl
117635
5-chloro-2-nitro-N-phenylaniline
C1=CC=C(C=C1)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-]
25781-92-4
C12H9ClN2O2
248.666
MFCD00007287
FPKHZBVGKMTUHB-UHFFFAOYSA-N
5-chloro-2-nitrodiphenylamine,benzenamine, 5-chloro-2-nitro-n-phenyl,5-chloro-2-nitrophenyl phenylamine,ghl.pd_mitscher_leg0.920,ksc203m3b,2-anilino-4-chloronitrobenzene,fpkhzbvgkmtuhb-uhfffaoysa,5-chloro-2-nitro-n-phenyl-benzenamine,benzenamine,5-chloro-2-nitro-n-phenyl
117635
5-chloro-2-nitro-N-phenylaniline
C1=CC=C(C=C1)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-]
Melting Point 108°C to 111°C
Quantity 5g
Formula Weight 248.67
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-nitrodiphenylamine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning