Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitrotoluene, 98%, Acros Organics

Click to view available options
Quantity:
25g
5367-28-2
C7H6ClNO2
171.58
MFCD00024579
NSMZCUAVEOTJDS-UHFFFAOYSA-N
5-chloro-2-nitrotoluene,toluene, 5-chloro-2-nitro,3-chloro-6-nitrotoluene,benzene, 4-chloro-2-methyl-1-nitro,2-nitro-5-chlorotoluene,benzene, 1-chloro-3-methyl-4-nitro-9ci,4-chloro-2-methyl-1-nitro-benzene,acmc-1am5r,5-chloro-2-nitro-toluene,ksc494a7d
79329
4-chloro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)Cl)[N+](=O)[O-]
5367-28-2
C7H6ClNO2
171.58
MFCD00024579
NSMZCUAVEOTJDS-UHFFFAOYSA-N
5-chloro-2-nitrotoluene,toluene, 5-chloro-2-nitro,3-chloro-6-nitrotoluene,benzene, 4-chloro-2-methyl-1-nitro,2-nitro-5-chlorotoluene,benzene, 1-chloro-3-methyl-4-nitro-9ci,4-chloro-2-methyl-1-nitro-benzene,acmc-1am5r,5-chloro-2-nitro-toluene,ksc494a7d
79329
4-chloro-2-methyl-1-nitrobenzene
CC1=C(C=CC(=C1)Cl)[N+](=O)[O-]
Infrared Spectrum Authentic
Refractive Index 1.5700 to 1.5720 (20°C, 589nm)
Packaging Glass bottle
Color Yellow
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(NO2)CH3
Beilstein 05, 327
Formula Weight 171.58
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 5-Chloro-2-nitrotoluene, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-355-3

RTECSNumber : XS9134000