Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-2-pentanone, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5891-21-4
C5H9ClO
120.58
MFCD00001008
XVRIEWDDMODMGA-UHFFFAOYSA-N
5-chloro-2-pentanone,2-pentanone, 5-chloro,1-chloro-4-pentanone,3-chloropropyl methyl ketone,2-pentanone 5-chloro,1-chloro-4-oxopentane,pubchem17207,3-acetylpropyl chloride,5-chloro-pentan-2-one,5-chloranylpentan-2-one
79993
5-chloropentan-2-one
CC(=O)CCCCl
5891-21-4
C5H9ClO
120.58
MFCD00001008
XVRIEWDDMODMGA-UHFFFAOYSA-N
5-chloro-2-pentanone,2-pentanone, 5-chloro,1-chloro-4-pentanone,3-chloropropyl methyl ketone,2-pentanone 5-chloro,1-chloro-4-oxopentane,pubchem17207,3-acetylpropyl chloride,5-chloro-pentan-2-one,5-chloranylpentan-2-one
79993
5-chloropentan-2-one
CC(=O)CCCCl
Boiling Point 172.0°C to 175.0°C
Infrared Spectrum Authentic
Density 1.0530g/mL
Color Brown to Black
Quantity 25g
Specific Gravity 1.053
Refractive Index 1.4370 to 1.4460
Packaging Glass bottle
Flash Point 35°C
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula Cl(CH2)3COCH3
Formula Weight 120.58
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 5-Chloro-2-pentanone
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 227-565-8