Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-phenylimidazole-4-carboxaldehyde, 97%, Thermo Scientific™

Catalog Number AAH3379003
Click to view available options
Quantity:
250 mg
1 g
60367-52-4
C10H7ClN2O
206.629
MFCD01935953
CCXUATWKEDQRMP-UHFFFAOYSA-N
5-chloro-2-phenyl-3h-imidazole-4-carbaldehyde, 4-chloro-2-phenyl-1h-imidazole-5-carbaldehyde, 5-chloro-2-phenyl-1h-imiadazole-4-carbaldehyde, 5-chloro-2-phenylimidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 5-chloro-2-phenyl, pubchem18833, ksc496q2r, chlorophenylimidazolecarbaldehyde, 2-phenyl-4-chloro-5-formylimidazole, 4-chloro-5-formyl-2-phenylimidazole
2763376
5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
C1=CC=C(C=C1)C2=NC(=C(N2)Cl)C=O
60367-52-4
C10H7ClN2O
206.629
MFCD01935953
CCXUATWKEDQRMP-UHFFFAOYSA-N
5-chloro-2-phenyl-3h-imidazole-4-carbaldehyde, 4-chloro-2-phenyl-1h-imidazole-5-carbaldehyde, 5-chloro-2-phenyl-1h-imiadazole-4-carbaldehyde, 5-chloro-2-phenylimidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 5-chloro-2-phenyl, pubchem18833, ksc496q2r, chlorophenylimidazolecarbaldehyde, 2-phenyl-4-chloro-5-formylimidazole, 4-chloro-5-formyl-2-phenylimidazole
2763376
5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
C1=CC=C(C=C1)C2=NC(=C(N2)Cl)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown-Orange or Pink
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 206.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chloro-2-phenylimidazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit