Learn More
Alfa Aesar™ 5-Chloro-2-(trifluoromethoxy)aniline, 97%
Manufacturer:  Alfa Aesar™ H32259MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH32259MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
326-64-7
211.57
UN2810
C7H5ClF3NO
MFCD06410919
5-Chloro-2-(trifluoromethoxy)aniline
Specifications
250mg
C7H5ClF3NO
MFCD06410919
5-Chloro-2-(trifluoromethoxy)aniline
326-64-7
211.57
UN2810
97%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.