Alfa Aesar™ 5-Chloro-2-(trifluoromethyl)phenylacetonitrile, 97%

Manufacturer: Alfa Aesar™ B2362906

Quantity 5g

 View more versions of this product

Catalog No. AAB2362906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261763-26-2
    Molecular Formula C9H5ClF3N
    Formula Weight 219.59
    MDL Number MFCD01631357
    UN Number UN3439
    Synonym 5-Chloro-2-(trifluoromethyl)benzyl cyanide
    Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetonitrile

    Specifications

    Formula Weight 219.59
    Melting Point 61° to 63°C
    Molecular Formula C9H5ClF3N
    MDL Number MFCD01631357
    CAS 261763-26-2
    UN Number UN3439
    Synonym 5-Chloro-2-(trifluoromethyl)benzyl cyanide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetonitrile
    Assay 97%

    GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit