Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-2-(trifluoromethyl)phenylacetonitrile, 97%

Click to view available options
Quantity:
1g
5g
261763-26-2
C9H5ClF3N
219.591
MFCD01631357
XCPSZOAJWIBKNE-UHFFFAOYSA-N
5-chloro-2-trifluoromethyl phenylacetonitrile,2-5-chloro-2-trifluoromethyl phenyl acetonitrile,5-chloro-2-trifluoromethyl phenyl acetonitrile,3-chloro-2-6-trifluoromethyl phenylacetonitrile,2-5-chloro-2-trifluoromethyl phenyl ethanenitrile,pubchem4286,acmc-20am4x,5-chloro-2-trifluoromethylbenzyl cyanide,5-chloro-2-trifluoromethyl benzyl cyanide
2778126
2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
C1=CC(=C(C=C1Cl)CC#N)C(F)(F)F
261763-26-2
C9H5ClF3N
219.591
MFCD01631357
XCPSZOAJWIBKNE-UHFFFAOYSA-N
5-chloro-2-trifluoromethyl phenylacetonitrile,2-5-chloro-2-trifluoromethyl phenyl acetonitrile,5-chloro-2-trifluoromethyl phenyl acetonitrile,3-chloro-2-6-trifluoromethyl phenylacetonitrile,2-5-chloro-2-trifluoromethyl phenyl ethanenitrile,pubchem4286,acmc-20am4x,5-chloro-2-trifluoromethylbenzyl cyanide,5-chloro-2-trifluoromethyl benzyl cyanide
2778126
2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
C1=CC(=C(C=C1Cl)CC#N)C(F)(F)F
Quantity 1g
Melting Point 61°C to 63°C
UN Number UN3439
Formula Weight 219.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-(trifluoromethyl)phenylacetonitrile

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.