Alfa Aesar™ 5-Chloro-3-methylbenzo[b]thiophene, 97%

Manufacturer: Alfa Aesar™ A1182714

Quantity 25g

 View more versions of this product

Catalog No. AAA1182714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19404-18-3
    Molecular Formula C9H7ClS
    Formula Weight 182.67
    MDL Number MFCD00052506
    Synonym 5-Chloro-3-methylthianaphthene
    Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene

    Specifications

    Formula Weight 182.67
    Molecular Formula C9H7ClS
    CAS 19404-18-3
    Synonym 5-Chloro-3-methylthianaphthene
    Melting Point 33° to 36°C
    Quantity 25g
    MDL Number MFCD00052506
    Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit