Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-3-methylbenzo[b]thiophene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
19404-18-3
C9H7ClS
182.67
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene,5-chloro-3-methylthianaphthene,5-chloro-3-methyl benzo b thiophene,5-chloro-3-methylbenzothiophene,4-methylthio phenol,zlchem 217,pubchem7372,acmc-209exr,maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
19404-18-3
C9H7ClS
182.67
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methylbenzo b thiophene,5-chloro-3-methylthianaphthene,5-chloro-3-methyl benzo b thiophene,5-chloro-3-methylbenzothiophene,4-methylthio phenol,zlchem 217,pubchem7372,acmc-209exr,maybridge1_005366
317317
5-chloro-3-methyl-1-benzothiophene
CC1=CSC2=C1C=C(C=C2)Cl
Melting Point 31.0°C to 32.0°C
Quantity 25g
Boiling Point 87.0°C (0.5 mmHg)
Assay Percent Range 96% min. (GC)
Formula Weight 182.67
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF