Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-3-phenyl-2,1-benzisoxazole, 98+%

Click to view available options
Quantity:
10g
50g
250g
719-64-2
C13H8ClNO
229.663
MFCD00014573
MUHJZJKVEQASGY-UHFFFAOYSA-N
5-chloro-3-phenylbenzo c isoxazole,5-chloro-3-phenylanthranil,5-chloro-3-phenyl-2,1-benzisoxazole,3-phenyl-5-chloroanthranil,pubchem8713,acmc-209okx,maybridge1_004397,2, 5-chloro-3-phenyl,5-chloro-3-phenyl-benzo c isoxazole,3-phenyl-5-chloro-2,1-benzisoxazole
347291
5-chloro-3-phenyl-2,1-benzoxazole
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl
719-64-2
C13H8ClNO
229.663
MFCD00014573
MUHJZJKVEQASGY-UHFFFAOYSA-N
5-chloro-3-phenylbenzo c isoxazole,5-chloro-3-phenylanthranil,5-chloro-3-phenyl-2,1-benzisoxazole,3-phenyl-5-chloroanthranil,pubchem8713,acmc-209okx,maybridge1_004397,2, 5-chloro-3-phenyl,5-chloro-3-phenyl-benzo c isoxazole,3-phenyl-5-chloro-2,1-benzisoxazole
347291
5-chloro-3-phenyl-2,1-benzoxazole
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl
Melting Point 114°C to 117°C
Quantity 10g
Formula Weight 229.67
Percent Purity ≥98%
Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA