Alfa Aesar™ 5-Chloro-4-azaindole-2-carboxylic acid, 98%

Manufacturer: Alfa Aesar™ H33628MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33628MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 800401-63-2
    Molecular Formula C8H5ClN2O2
    Formula Weight 196.59
    MDL Number MFCD18261123
    Synonym 5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid
    Chemical Name or Material 5-Chloro-4-azaindole-2-carboxylic acid

    Specifications

    Formula Weight 196.59
    Molecular Formula C8H5ClN2O2
    MDL Number MFCD18261123
    Synonym 5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈295°C
    Quantity 250mg
    CAS 800401-63-2
    Chemical Name or Material 5-Chloro-4-azaindole-2-carboxylic acid
    Assay 98%

    Description

    Applications
    Used for pharmaceutical and cytokine signaling intermediates.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit