Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-4-fluoro-2-nitroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
104222-34-6
C6H4ClFN2O2
190.56
MFCD00052698
VRJKEIWZSOHDOH-UHFFFAOYSA-N
2-nitro-4-fluoro-5-chloroaniline,benzenamine, 5-chloro-4-fluoro-2-nitro,benzenamine,5-chloro-4-fluoro-2-nitro,5-chloro-4-fluoro-2-nitrophenylamine,pubchem8452,acmc-2098bg,timtec-bb sbb003647,buttpark 45\03-11,3-chloro-4-fluoro-6-nitroaniline,5-chloro-4-fluoro-2-nitro-aniline
517835
5-chloro-4-fluoro-2-nitroaniline
C1=C(C(=CC(=C1Cl)F)[N+](=O)[O-])N
104222-34-6
C6H4ClFN2O2
190.56
MFCD00052698
VRJKEIWZSOHDOH-UHFFFAOYSA-N
2-nitro-4-fluoro-5-chloroaniline,benzenamine, 5-chloro-4-fluoro-2-nitro,benzenamine,5-chloro-4-fluoro-2-nitro,5-chloro-4-fluoro-2-nitrophenylamine,pubchem8452,acmc-2098bg,timtec-bb sbb003647,buttpark 45\03-11,3-chloro-4-fluoro-6-nitroaniline,5-chloro-4-fluoro-2-nitro-aniline
517835
5-chloro-4-fluoro-2-nitroaniline
C1=C(C(=CC(=C1Cl)F)[N+](=O)[O-])N
Melting Point 142°C to 145°C
Quantity 1g
Packaging Glass bottle
Color Orange
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 190.56
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Chloro-4-fluoro-2-nitroaniline
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning