Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-4-methyl-2-nitroaniline, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7149-80-6
C7H7ClN2O2
186.6
MFCD00034066
BBWHBSDZHSQECL-UHFFFAOYSA-N
5-chloro-4-methyl-2-nitro-phenylamine,benzenamine, 5-chloro-4-methyl-2-nitro,5-chloro-4-methyl-2-nitrophenylamine,5-chloro-4-methyl-2-nitroaniline,acmc-209oix,4-amino-2-chloro-5-nitrotoluene,zerenex e/9102011,5-chloro-4-methyl-2-nitro-aniline,5-chloro-2-nitro-p-toluidine,benzenamine,5-chloro-4-methyl-2-nitro
251681
5-chloro-4-methyl-2-nitroaniline
CC1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
7149-80-6
C7H7ClN2O2
186.6
MFCD00034066
BBWHBSDZHSQECL-UHFFFAOYSA-N
5-chloro-4-methyl-2-nitro-phenylamine,benzenamine, 5-chloro-4-methyl-2-nitro,5-chloro-4-methyl-2-nitrophenylamine,5-chloro-4-methyl-2-nitroaniline,acmc-209oix,4-amino-2-chloro-5-nitrotoluene,zerenex e/9102011,5-chloro-4-methyl-2-nitro-aniline,5-chloro-2-nitro-p-toluidine,benzenamine,5-chloro-4-methyl-2-nitro
251681
5-chloro-4-methyl-2-nitroaniline
CC1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
Melting Point 159°C to 163°C
Quantity 1g
Packaging Glass bottle
Color Orange to Brown
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 186.6
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 5-chloro-4-methyl-2-nitroaniline, 96%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning