Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Chloro-7-azaindole, 95%

Click to view available options
Quantity:
1g
5g
866546-07-8
C7H5ClN2
152.581
MFCD06659676
MFZQJIKENSPRSJ-UHFFFAOYSA-N
5-chloro-1h-pyrrolo 2,3-b pyridine,5-chloro-7-azaindole,1h-pyrrolo 2,3-b pyridine, 5-chloro,pubchem14704,ksc494e6r,5-chloropyrrolo 2,3-b pyridine,5-chloro-1h-pyrrolo 2,3-b pyridin,5-chloro-1h-pyrrolo 2,3-b pyridine
24229213
5-chloro-1H-pyrrolo[2,3-b]pyridine
C1=CNC2=NC=C(C=C21)Cl
866546-07-8
C7H5ClN2
152.581
MFCD06659676
MFZQJIKENSPRSJ-UHFFFAOYSA-N
5-chloro-1h-pyrrolo 2,3-b pyridine,5-chloro-7-azaindole,1h-pyrrolo 2,3-b pyridine, 5-chloro,pubchem14704,ksc494e6r,5-chloropyrrolo 2,3-b pyridine,5-chloro-1h-pyrrolo 2,3-b pyridin,5-chloro-1h-pyrrolo 2,3-b pyridine
24229213
5-chloro-1H-pyrrolo[2,3-b]pyridine
C1=CNC2=NC=C(C=C21)Cl
Quantity 1g
Color Brown to White
Melting Point 161°C to 165°C
UN Number UN2811
Beilstein 11085807
Formula Weight 152.58
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 5-Chloro-7-azaindole

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.