Alfa Aesar™ 5-Chloro-8-hydroxy-7-iodoquinoline, 98%

Manufacturer: Alfa Aesar™ A1113730

Quantity 250g

 View more versions of this product

Catalog No. AAA1113730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 130-26-7
    Molecular Formula C9H5ClINO
    Formula Weight 305.49
    MDL Number MFCD00006787
    Beilstein 153637
    Merck Index 14,5031
    Synonym Clioqinol
    Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline

    Specifications

    Formula Weight 305.49
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 130-26-7
    Beilstein 153637
    Merck Index 14,5031
    Quantity 250g
    TSCA TSCA
    Molecular Formula C9H5ClINO
    Synonym Clioqinol
    Health Hazard 1 Danger
    Melting Point ≈175°C (dec.)
    MDL Number MFCD00006787
    Health Hazard 2 GHS H Statement
    H334-H361-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of damaging fertility or the unborn child.
    May cause an allergic skin reaction.
    RTECS Number VC5075000
    Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline
    Assay 98%

    Description

    Applications
    Clioquinol is a metal chelating ligand which displays protective effects in various neurodegenerative states as well as being a component in antifungal and antiprotozoal agents. Clioquinol rescues lead-induced impairments of synaptic plasticity in an animal models

    Solubility
    Soluble in DMSO (>25 mg/ml), boiling alcohol ((1:43)), methanol, and chloroform ((1:120)).

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H334-H361-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of damaging fertility or the unborn child.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit